missing translation for 'onlineSavingsMsg'
Learn More

Turbidity QC Sample, Drinking Water Standard, SPEX CertiPrep™

Turbidity QC Samples for drinking water analysis, 15 mL, SPEX CertiPrep™

Brand:  SPEX Certiprep 699

Product Code. 15628215

  • 163.00 € / 15mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
This item is not returnable. View return policy

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

  • Turbidity QC Sample
  • QC Samples for drinking water
  • 15 mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Turbidity QC Sample
Triple-leached LDPE bottle
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Yields up to 1L after dilution
15 mL
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product intended for laboratory use only