missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TurboFect™ Transfection Reagent

Deliver DNA plasmids and expression vectors into eukaryotic cells with this highly efficient, easy-to-use, non-immunogenic transfection reagent.

Brand:  Thermo Scientific™ R0531

265.24 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.


 View more versions of this product

Product Code. 15325016

  • 290.00 € / 1mL

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

The Thermo Scientific™ TurboFect in vitro Transfection Reagent is a highly efficient, easy-to-use, non-immunogenic transfection reagent for delivery of DNA plasmids and expression vectors into eukaryotic cells.

The polymer forms compact, stable, positively charged complexes with DNA. These complexes protect DNA from degradation and facilitate gene delivery in vivo. Reagent causes no detectable inflamatory response and is suitable for DNA administration via various routes, including intravenous, intraperitoneal, intratumoral injection, etc.
  • No detectable inflamatory response
  • Gene delivery via multiple routes of administration
  • Reproducible results
  • Functionally tested in mice and rats

Recommended for:

DNA transfection

siRNA-DNA co-transfection

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Transfection Reagent
Chemically Defined
6-Well Plate, 12-Well Plate, 24-Well Plate, 48-Well Plate, 96-Well Plate, Flasks
TurboFect
1 mL
Primary Cell, Endothelial Cell, Epithelial Cell, Myeloma Cell, Cell Lines
Chemical Transfection
Transfection
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ TurboFect™ Transfection Reagent > 1 mL

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis

For Research Use Only. Not for use in diagnostic procedures.