missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TV-1665 Vertical Rotor

Spin samples at 65,000rpm with the exceptional TV-1665 vertical rotor.

Brand:  Thermo Scientific™ 11672

Product Code. 12161640

  • 28840.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Spin samples at 65,000rpm using the exceptional Thermo Scientific™ TV-1665 Vertical Rotor.

Compatible with:

Thermo Scientific Sorvall WX 80, 90, 100 Ultra Series Floor Model Centrifuge

Recommended for:

Ideal for gradient applications.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

TV-1665 Vertical Rotor
Vertical Rotor
414,357 x g
87.8 mm
9.8
13 mm
Thermo Scientific™ Sorvall™ WX ultraCentrifuges
383,209 x g
Titanium
16 x 6 mL
65,000 rpm
74.6 mm
66 mm
19.9 lb. (9.07 kg)
352,062 x g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.