missing translation for 'onlineSavingsMsg'
Learn More

UVP TW-26 Transilluminator

Back-lit, white light transilluminator with uniform light diffusion. UVP Ultra Violet Product™ TW-26 Transilluminator can be used to examine autoradiographs, X-ray film, and microtiter plates.

Brand:  UVP 95-0208-02

Product Code. 12621732

  • 1205.00 € / Each
Estimated Shipment: 07-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Plexiglass and glass surfaces are double-safety sealed into the unit for durability, providing a scratch- and chemical-resistant work area
  • Uniform light diffusion
  • Two-year warranty
Specifications

Specifications

Single
580/630 nm
White Light Transilluminator
210 x 260 mm
230 V 50 Hz
TW-26
50 Hz
White
108 mm
8 w
241 x 337 x 108mm
210 x 260 mm
230 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers