missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Tween™ 20 Surfact-Amps™ Detergent Solution

Thermo Scientific™ Surfact-Amps 20 Detergent Solution is highly-purified Tween-20 detergent stabilized and supplied as a 10% solution in 10mL glass ampules.

Brand:  Thermo Scientific™ 28321

 View more versions of this product

Product Code. 11368311

  • 2205.00 € / 1000mL
Estimated Shipment: 27-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Tween-20 Detergent Surfact-Amps Solution is highly-purified Tween-20 detergent stabilized and supplied as a 10% solution in 10mL glass ampules.

This is an easy-to-use 10% (w/v) solution of purified Tween-20 detergent for use in routine and high-demand protein research methods and molecular biology techniques. Tween-20 is a nonionic polyoxyethylene surfactant that is most frequently added to PBS or TBS wash buffers for ELISA, Western blotting and other immunoassay methods.

Surfact-Amps™ Detergent Solutions (10% w/v) provide unsurpassed purity, quality and stability. Unlike neat detergents, which are extremely viscous, Surfact-Amps 10% Solutions are easy to pipette and accurately dispense. The surfactant solutions are carefully prepared and packaged under nitrogen in glass ampules, ensuring their stability and eliminating the accumulation of peroxides and degradation products.

Highlights:

Tween 20 – a popular nonionic detergent for use in immunoassay wash buffers

Accurate – precise 10% detergent solution in ultrapure water

Easy-to-use – solution is simple to dispense and dilute for use

Exceptionally pure – less than 1.0μeq/ml peroxides and carbonyls

Stable – packed in glass ampules under inert nitrogen gas

Detergent Properties:

  • Molecular Weight: 1228
  • Detergent Class: Nonionic
  • Aggregation Number: Unknown
  • Micelle Molecular Weight: Unknown
  • Critical Micelle Concentration (CMC): 0.06mM (0.0074%, w/v)
  • Cloud Point: 95°C
  • Dialyzable: No

 

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store in a cool, dry, and well-ventilated area
Surfact-Amps™, Tween™
1 L
Liquid
Detergent Solution
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.