missing translation for 'onlineSavingsMsg'
Learn More

TWEEN 20, MP Biomedicals™

Product Code. 11417160
Click to view available options
Quantity:
500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

TWEEN 20, Molecular biology reagent, Viscous pale yellow liquid

  • TWEEN 20 is a nonionic detergent widely used in biochemical applications, such as emulsifying agents for the preparation of stable oil-inwater emulsions.
  • It has been used in preextraction of membranes to remove peripheral proteins.
  • TWEEN 20 has been used as a blocking agent for membrane based immunoassays at a typical concentration of 0.05%.
  • TWEEN 20 can be used for lysing mammalian cells at a concentration of 0.05 to 0.5%. TWEEN 20 is a polyoxyethylene sorbitol ester.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9005-64-5
Molecular Formula (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6
Molecular Weight (g/mol) 522.68
MDL Number MFCD00165986
InChI Key HMFKFHLTUCJZJO-UHFFFAOYNA-N
Synonym Polyethyleneglycol sorbitan monolaurate, Polyoxyethylenesorbitan monolaurate, Polysorbate 20, Polyoxyethylene (20) Sorbitan Monolaurate
IUPAC Name 2-{2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl dodecanoate
SMILES CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO

Specifications

Color Pale Yellow
Physical Form Liquid
Boiling Point >100°C (literature)
Density 1.09 - 1.12(25°C)
Quantity 500 mL
Flash Point >230°F/ >110°C (Lit.)
Refractive Index < 1.33 hPa (< 1.00 mmHg, Lit.)
Specific Gravity 1.08 - 1.13
Viscosity Brookfield Viscosity: 370-430 cps (25°C, neat, Lit.)
Water ≤3%
Solubility Information Soluble in Water(100mg/mL).
Grade Molecular Biology
Show More Show Less
compliance-icons
Product Identifier
  • Tween 20, molecular biology reagent
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.