missing translation for 'onlineSavingsMsg'
Learn More

Tween 20™, Ultrapure, Thermo Scientific Chemicals

44.90 € - 114.48 €

Chemical Identifiers

CAS 9005-64-5
Molecular Formula (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6
Molecular Weight (g/mol) 522.68
MDL Number MFCD00165986
InChI Key HMFKFHLTUCJZJO-UHFFFAOYNA-N
Synonym Polyoxyethylene sorbitan monolaurate
PubChem CID 443314
SMILES CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15825398
View Documents Promotion Details
Thermo Scientific
J20605.AP
500 mL Plastic bottle
44.90 €
500mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15895178
View Documents Promotion Details
Thermo Scientific
J20605.LO
1 gal. Plastic bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Chemical Identifiers

9005-64-5
522.68
HMFKFHLTUCJZJO-UHFFFAOYNA-N
443314
(C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6
MFCD00165986
Polyoxyethylene sorbitan monolaurate
CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO

Specifications

9005-64-5
>100°C
(C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6
Polyoxyethylene sorbitan monolaurate
HMFKFHLTUCJZJO-UHFFFAOYNA-N
2-[2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy]ethyl dodecanoate
443314
1.105
Liquid
1.105
MFCD00165986
>110°C
CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
522.68
Ultra Pure
Tween 20™
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 500-018-3

missing translation for 'rtecsNumber' : TR7400000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only