missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tween 20 washing buffer, 0.5% in PBS (10X)

60.50 € - 149.00 €

Specifications

Product Type Buffer
Form Liquid
Concentration 0.5% in PBS (10X)
Content And Storage Ambient temperatures
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15485789
View Documents
Thermo Scientific Chemicals
J63596.AP
500 mL
60.50 €
500mL
Please to purchase this item. Need a web account? Register with us today!
15495789
View Documents
Thermo Scientific Chemicals
J63596.K2
1 L
88.94 €
1 L
Please to purchase this item. Need a web account? Register with us today!
15405799
View Documents
Thermo Scientific Chemicals
J63596.K3
2 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Tween 20 washing buffer, 0.5% in PBS (10X) is used as a wash solution for western blot membranes and microtiter plate wells in ELISA assays. It acts as a pH stabilizer in optimal formulation, salts and detergents designed to effectively remove excess material from membranes. It finds application as laboratory detergents as well as in biomedical experiments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Buffer
Liquid
0.5% in PBS (10X)
Ambient temperatures
Videos
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only