Learn More
Thermo Scientific Chemicals Tween 20 washing buffer, 1% in PBS (10X)
66.03 € - 154.77 €
Specifications
Product Type | Buffer |
---|---|
Form | Viscous Liquid |
Concentration | 1% in PBS (10X) |
Content And Storage | Keep cold |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15435579
|
Thermo Scientific Chemicals
J63314.K2 |
1 L |
88.24 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15425579
|
Thermo Scientific Chemicals
J63314.AP |
500 mL |
66.03 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15445579
|
Thermo Scientific Chemicals
J63314.K3 |
2 L |
154.77 €
2 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tween-20 is a non-ionic surfactant that is useful for solubilizing membrane proteins during isolation. Tween-20 is also added to buffers for immunohistochemistry. Tween-20 may also help to prevent non-specific antibody binding and function as a blocking agent in immunoblotting assays. It has been used in preextraction of membranes to remove peripheral proteins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Buffer | |
Viscous Liquid | |
1% in PBS (10X) | |
Keep cold |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.