Learn More
Thermo Scientific™ Tween™ 80 Surfact-Amps™ Detergent Solution
Thermo Scientific™ Surfact-Amps 80 Detergent Solution is highly-purified Tween-80 detergent stabilized and supplied as a 10% solution in 10mL glass ampules.
186.00 € - 2305.00 €
Specifications
Content And Storage | Store in a cool, dry, and well-ventilated area |
---|---|
Form | Liquid |
Product Line | Surfact-Amps™, Tween™ |
Reagent Type | Detergent Solution |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
13494259
|
Thermo Scientific™
28329 |
50 mL |
186.00 €
50mL |
In Stock
Log in to see stock. |
|||||
11871455
|
Thermo Scientific™
28328 |
6 x 10 mL |
206.00 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
13404269
|
Thermo Scientific™
28230 |
500 mL |
2305.00 €
500mL |
Estimated Shipment: 09-01-2025
Log in to see stock. |
|||||
Description
Thermo Scientific™ Tween-80 Detergent Surfact-Amps™ Solution is highly-purified Tween-80 detergent stabilized and supplied as a 10% solution in 10mL glass ampules.
This is an easy-to-use 10% (w/v) solution of purified Tween-80 detergent for use in routine and high-demand protein research methods and molecular biology techniques. Tween-80 is a nonionic polyoxyethylene surfactant that is most frequently added to PBS or TBS wash buffers for ELISA, Western blotting and other immunoassay methods.
Surfact-Amps™ Detergent Solutions (10% w/v) provide unsurpassed purity, quality and stability. Unlike neat detergents, which are extremely viscous, Surfact-Amps 10% Solutions are easy to pipette and accurately dispense. The surfactant solutions are carefully prepared and packaged under nitrogen in glass ampules, ensuring their stability and eliminating the accumulation of peroxides and degradation products.
Highlights:
Tween 80 – a nonionic detergent for use in wash buffers
Accurate – precise 10% detergent solution in ultrapure water
Easy-to-use – solution is simple to dispense and dilute for use
Exceptionally pure – less than 1.0μeq/ml peroxides and carbonyls
Stable – packed in glass ampules under inert nitrogen gas
Detergent Properties:
- Molecular Weight: 1310
- Detergent Class: Nonionic
- Aggregation Number: 60
- Micelle Molecular Weight: 76,000
- Critical Micelle Concentration (CMC): 0.012mM (0.0016%, w/v)
- Cloud Point: Unknown
- Dialyzable: No
Specifications
Store in a cool, dry, and well-ventilated area | |
Liquid | |
Surfact-Amps™, Tween™ | |
Detergent Solution |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.