Learn More
Eppendorf™ twin.tec™ PCR Plates BioBased
BIOPHARMAGREENER_CHOICE
Discover the new twin.tec™ PCR Plates BioBased made of bio-based polymer (mass balance approach). These plates allow you to significantly reduce your consumable-related carbon footprint while maintaining identical technical performance.
Brand:  Eppendorf™ 0030531078
280.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 18008804
  • 285.00 € / Pack of 25
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Reduction of consumable-related carbon footprint in the lab
  • Laser-engraved lot number and expiration date on each single plate to improve traceability in your laboratory processes
  • Unique laser-engraved optical guiding grid and OptiTrack matrix for quick orientation when pipetting manually
  • One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performance
  • Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample
  • Exceptionally solid, torque- and heat-resistant polycarbonate frame: No warping, bending or breaking during heat incubation or centrifugation
  • Raised well rims for effective sealing, also reduces risk of cross-contamination
  • Batch-tested and certified free of DNA, DNAase, RNase and PCR inhibitors (PCR clean)
  • Available with barcode (upon request)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Custom Barcoding Available
Eppendorf
BioBased LoBind PCR Plates
Polypropylene, Polycarbonate
Non-sterile
25 Pk.
LoBind
Colorless
Skirted
384
Conical Bottom (V-well)
PCR Ready
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Eppendorf™ twin.tec™ PCR Plates BioBased > Product Type: PCR Plates BioBased LoBind™, No. of Wells: 384, Color: Colorless, Skirt Style: Skirted