Learn More
Eppendorf™ twin.tec™ real-time qPCR Plates, 96-well - qPCR Plates
GREENER_CHOICE
Eppendorf twin.tec™ real-time PCR plates, designed for low-volume qPCR, utilize white wells to enhance fluorescent light reflection, improving sensitivity and reproducibility in assays for reliable results.
Brand:  Eppendorf™ 0030129636
 View more versions of this product
Product Code. 15685367
Quantity Price
1 76.90 € / Pack of 10
5 71.06 € / Pack of 10
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • White wells for better reflection.
  • High mechanical stability.
  • Effective sealing by raised well rims.
  • Skirted and semi-skirted frames.
  • Optimal heat transfer due to reduced wall thickness.
  • Autoclavable (121°C, 20 min.).
  • Available in lot-certified PCR clean and Eppendorf Forensic DNA Grade for tested purity.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Standard
Autoclavable
White
Skirted
150 μL
Non-sterile
6000 x g
10 Pk.
White
8 x 12
10 x Individually Blister Wrapped
PCR Plate
Polypropylene, Polycarbonate
96
Real-time PCR
Forensic DNA Grade (Non-sterile)
Yes