missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ twin.tec™ real-time qPCR Plates, 96-well - qPCR Plates
GREENER_CHOICE

Eppendorf twin.tec™ real-time PCR plates, designed for low-volume qPCR, utilize white wells to enhance fluorescent light reflection, improving sensitivity and reproducibility in assays for reliable results.

Brand:  Eppendorf™ 0030129644

75.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 15695367

  • 75.85 € / Pack of 10
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • White wells for better reflection.
  • High mechanical stability.
  • Effective sealing by raised well rims.
  • Skirted and semi-skirted frames.
  • Optimal heat transfer due to reduced wall thickness.
  • Autoclavable (121°C, 20 min.).
  • Available in lot-certified PCR clean and Eppendorf Forensic DNA Grade for tested purity.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Standard
Autoclavable
White
Semi-skirted
250 μL
Non-sterile
6000 x g
10 Pk.
White
8 x 12
10 x Individually Blister Wrapped
PCR Plate
Polypropylene, Polycarbonate
96
Real-time PCR
Forensic DNA Grade (Non-sterile)
Yes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Eppendorf™ twin.tec™ real-time qPCR Plates, 96-well - qPCR Plates > Sterility: Forensic DNA Grade (Non-sterile), Well Volume: 250 μL, Skirt Style: Semi-Skirted, Color: White, Packaging: 10 x Individually Blister Wrapped

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.