Learn More
Fisherbrand Twirl'EM™ Clear Polyethylene Sampling Bags
Product Code. p-8010771
Click to view available options
Capacity (Metric):
150 mL
650 mL
800 mL
1060 mL
1240 mL
2250 mL
4000 mL
Unit Size:
Pack of 1000
Product Code. 11582773
Brand: Fisherbrand EFL1012
Only null left
Add to basket
Add to basket
Collect solid, semi-solid or liquid samples in the laboratory using Twirl'EM™ Clear Polyethylene Sampling Bags. The bags feature a twirl round metal wire closure system with safety tabs to provide a leakproof seal.
  • Sterile
  • Clear polyethylene
  • Without side seals, opening at width
  • Pull tabs to open bag without touching edges
  • Twirl round metal wire closure system with safety tabs
Specifications
Capacity (Metric) 2250 mL
Color Clear
Material Polyethylene
Sterility Sterile
Quantity 1000 Pk.