Learn More
LABPLAS Twirl'Em Sample Bags
Twirl'em sample bags are safe, flexible, contaminant-free and easy to use.
Brand:  LABPLAS EFR-1015S-FS
 View more versions of this product
Product Code. 17950215
  • 1015.00 € / Pack of 1000
Explore more special offers
Description
Description
  • Twirl'Em sampling bags
  • Safety tabs
  • Clear, with writing area for easy labeling
  • Closure with 1 round wire and 1 flat wire
  • Available in various sizes
Specifications
Specifications
Sample Bag
Clear with White Block
Sterile
Yes
254 x 381 mm
254 mm
381 mm
Not Autoclavable
Single-use
101.6 μm
4 mil.
10 in.
15 in.