Learn More
Lenz Laborglasintrumente Two-Neck Round-Bottom Flasks, Side Neck Parallel
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 03204473
 View more versions of this product
Product Code. 10385921
  • 132.00 € / Each
Estimated Shipment: 06-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Distillation flasks
  • With parallel side neck
  • With conical ground joints
  • With inscription label
  • Material: DURAN™
Specifications
Specifications
2 L
Duran Glass
Side Neck NS 29/32
166 mm