Learn More
Thermo Scientific™ TX-200 Swinging Bucket Rotor
Accommodate a wide range of processing volumes with the TX-200 4 x 180mL Swinging Bucket Rotor.
Brand:  Thermo Scientific™ 75003687
144.22 EUR valid until 2024-12-31
Use promo code "21988" to get your promotional price.
 View more versions of this product
Product Code. 11765008
  • 158.79 € / Pack of 4
Explore more special offers
Description
Description
Accommodate a wide range of processing volumes with the Thermo Scientific™ TX-200 4 x 180mL Swinging Bucket Rotor.
Ordering Information:
Round buckets (75-003-659), biocontainment lids (75-003-660) and replacement O-rings (75-003-687) are sold separately. A variety of adapters are also available. Contact your sales representative for more information.
Compatible with:
Thermo Scientific Sorvall™ ST16, SL16, Sorvall Legend™ X1, Multifuge™ X1, Megafuge™ 16 Centrifuge Series
Recommended for:
Cell culture, blood separation, body fluids processing, microbiology, proteomics, genomics and molecular biology
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Replacement O-rings for Lids
Replacement O-ring
Thermo Scientific Sorvall Legend X1, Sorvall ST 16, Multifuge X1, Megafuge 16 and SL 16 Centrifuges
Yes