Learn More
Thermo Scientific™ TX-200 Swinging Bucket Rotor
Accommodate a wide range of processing volumes with the TX-200 4 x 180mL Swinging Bucket Rotor.
Brand: Thermo Scientific™ 75003659
844.47 EUR valid until 2025-12-31
Use promo code "24378" to get your promotional price.
Description
Accommodate a wide range of processing volumes with the Thermo Scientific™ TX-200 4 x 180mL Swinging Bucket Rotor.
Ordering Information:Round buckets (75-003-659), biocontainment lids (75-003-660) and replacement O-rings (75-003-687) are sold separately. A variety of adapters are also available. Contact your sales representative for more information.
Compatible with:
Thermo Scientific Sorvall™ ST16, SL16, Sorvall Legend™ X1, Multifuge™ X1, Megafuge™ 16 Centrifuge Series
Recommended for:
Cell culture, blood separation, body fluids processing, microbiology, proteomics, genomics and molecular biology
Specifications
Round Buckets for TX-200 Rotor (4) | |
Swinging Bucket Rotor | |
Thermo Scientific Sorvall Legend X1, Sorvall ST 16, Multifuge X1, Megafuge 16 and SL 16 Centrifuges | |
Yes |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.