missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tychem 2000 C
Shop All DuPont Products
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Unit Size:
Each
Pack of 25
Description
- Hooded coverall with elastic face opening
- Adhesive chin flap for tight fit around respirator
- Stitched and over-taped seams
- Souble self-adhesive zipper flap
- Elastic cuffs, ankles and waist
- Thumb loops prevent sleeves from riding up
- 100% particle tight fabric
- Excellent barrier against a broad range of concentrated inorganic chemicals and biological hazards
- Antistatic treatment (inside)
Specifications
Specifications
| Product Type | Hooded Coveralls |
| Size | X-Large |
| Material | Tychem |
| Color | Yellow |
| Waist Style | Elastic |
| Ankle Style | Elastic |
| Cuff Style | Thumb Loop |
| Neck Style | Collared |
| Hood Type | Elastic |
| Closure Type | Double Zipper with Self-adhesive Flap |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Übermitteln Sie eine inhaltliche Korrektur