Learn More
DuPont™ Tychem 2000 C
Bright yellow color provides high visibility. Lightweight and durable fabric and a polymeric barrier coating offers good permeation barrier protection against a wide range of inorganic chemicals and biological hazards (even under pressure).
Brand:  DuPont™ D13395560
20.00 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 12932159
  • 27.15 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Hooded coverall with elastic face opening
  • Adhesive chin flap for tight fit around respirator
  • Stitched and over-taped seams
  • Souble self-adhesive zipper flap
  • Elastic cuffs, ankles and waist
  • Thumb loops prevent sleeves from riding up
  • 100% particle tight fabric
  • Excellent barrier against a broad range of concentrated inorganic chemicals and biological hazards
  • Antistatic treatment (inside)
Order Info
dimSize
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Hooded Coveralls
Tychem
Elastic
Thumb Loop
Elastic
Stitched, Taped
Chemicals
Tyvek™ C Fabric
Tychem™ C is used for splash or pressurized splash protection in a variety of industrial environments, including pulp and paper manufacturing, food processing, chemical processing and pharmaceutical manufacturing
2X-Large
Yellow
Elastic
Hooded
Double Zipper with Self-adhesive Flap
Category III, Type 3-B, 4-B, 5-B and 6-B
EN 14126, EN 1073-2, EN 1149-5
116 to 124 cm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products