missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tychem™ 6000 F FaceSeal
Click to view available options
Size:
Small
Medium
Large
2X-Large
3X-Large
4X-Large
5X-Large
Description
- Made from a lightweight and durable fabric (< 500 g per garment) consisting of a proprietary barrier film laminated to a heavy duty Tyvek™ substrate
- Offer excellent chemical permeation protection to an extensive range of chemicals, helping to protect personnel against numerous toxic industrial organic chemicals, highly concentrated inorganic chemicals (even under pressure), particulates, biohazards, and certain chemical warfare agents
- Certified according to Regulation (EU) 2016/425
- Chemical protective clothing, Category III, Type 3-B, 4-B, 5-B and 6-B
- EN 14126 (barrier to infective agents), EN 1073-2 (protection against radioactive contamination)
- Antistatic treatment (EN 1149-5) on inside
- Stitched and over-taped seams with barrier tape for protection and strength
- Back entry with zipper, double zipper flap with snaps and hook and loop closure
- Attached undergloves
- Used for a broad range of applications from chemical spill clean-up, emergency response, to military and petrochemical applications
Specifications
Specifications
Color | Gray |
Size | Large |
Ankle Style | Attached socks |
Closure Type | Rear-entry zipper with double flap, snaps and hook-and-loop seal |
Cuff Style | Attached gloves |
Disposable | Single-use |
Gender | Unisex |
Neck Style | Hooded |
Quantity | 1/Ea. |
Seam Type | Stitched, Taped |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'