Learn More
DuPont™ Tychem™ 6000 F FaceSeal
Tychem™ 6000 F FaceSeal is a hooded coverall that includes a rubber seal for a tight fit.
136.00 € - 153.00 €
Specifications
Color Gray
Ankle Style Attached socks
Closure Type Rear-entry zipper with double flap, snaps and hook-and-loop seal
Cuff Style Attached gloves
Disposable Single-use
View More Specs
Products 7
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
16328555
View Documents
DuPont™
D15490341
Small
136.00 €
Each
Estimated Shipment: 21-01-2025
to see stock.
Add to basket
16348555
View Documents
DuPont™
D15490481
Medium
136.00 €
Each
Estimated Shipment: 17-01-2025
to see stock.
Add to basket
16338555
View Documents
DuPont™
D15490467
Large
136.00 €
Each
Estimated Shipment: 17-01-2025
to see stock.
Add to basket
16358555
View Documents
DuPont™
D15490598
2X-Large
136.00 €
Each
Estimated Shipment: 17-01-2025
to see stock.
Add to basket
16368555
View Documents
DuPont™
D15490650
3X-Large
142.00 €
Each
Estimated Shipment: 17-01-2025
to see stock.
Add to basket
16378555
View Documents
DuPont™
D15490706
4X-Large
147.00 €
Each
Estimated Shipment: 05-03-2025
to see stock.
Add to basket
16388555
View Documents
DuPont™
D15490762
5X-Large
153.00 €
Each
Estimated Shipment: 05-03-2025
to see stock.
Add to basket
Description
Description
  • Made from a lightweight and durable fabric (< 500 g per garment) consisting of a proprietary barrier film laminated to a heavy duty Tyvek™ substrate
  • Offer excellent chemical permeation protection to an extensive range of chemicals, helping to protect personnel against numerous toxic industrial organic chemicals, highly concentrated inorganic chemicals (even under pressure), particulates, biohazards, and certain chemical warfare agents
  • Certified according to Regulation (EU) 2016/425
  • Chemical protective clothing, Category III, Type 3-B, 4-B, 5-B and 6-B
  • EN 14126 (barrier to infective agents), EN 1073-2 (protection against radioactive contamination)
  • Antistatic treatment (EN 1149-5) on inside
  • Stitched and over-taped seams with barrier tape for protection and strength
  • Back entry with zipper, double zipper flap with snaps and hook and loop closure
  • Attached undergloves
  • Used for a broad range of applications from chemical spill clean-up, emergency response, to military and petrochemical applications
Specifications
Specifications
Gray
Rear-entry zipper with double flap, snaps and hook-and-loop seal
Single-use
Hooded
Stitched, Taped
Elastic
Attached socks
Attached gloves
Unisex
1/Ea.
Non-sterile