Learn More
DuPont™ Tychem™ 6000 F Orange Hooded Coverall
GREENER_CHOICE
Product Code. 12633680
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Offering excellent chemical permeation protection to an extensive range of chemicals. DuPont™ Tychem™ 6000 F Orange Hooded Coverall are made from a lightweight and durable fabric consisting of a proprietary barrier film-laminated to a heavy duty Tyvek substrate.
  • Available in grey for discretionary needs
  • Elasticated face, wrists, waist and ankles for optimal fit
  • Thumb loops prevent sleeves from riding up
  • Self-adhesive chin flap for tight seal of suit to face masks
  • Self-adhesive double zipper flap closure system
  • Stitched and over-taped seams with barrier tape for protection and strength
  • Protects personnel against numerous toxic industrial organic chemicals, highly concentrated inorganic chemicals (even under pressure), particulates, radioactive contamination, biohazards and certain chemical warfare agents
  • Used for a broad range of applications from chemical spill clean-up, emergency response to military and petrochemical applications
TRUSTED_SUSTAINABILITY
Specifications
Product Type Hooded Coveralls
Size 2X-Large
Material Tychem
Color Orange
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type Elastic
Closure Type Double Zipper with Self-adhesive Flap
Seam Type Stitched, Taped
Protection Class Category III, Type 3-B, 4-B, 5-B and 6-B
Resistance Chemicals
Collar Type Self-adhesive chin flap
Certifications/Compliance (EU) 2016/425, EN 14126, EN 1073-2, EN 1149-5
Size (Metric) Chest 116 to 124 cm
For Use With (Application) For a broad range of applications from chemical spill clean-up, emergency response, to military and petrochemical applications