Learn More
DuPont™ Tychem™ 6000 FR ThermoPro Product Code.: 15868267
DuPont™ Tychem™ 6000 FR ThermoPro
Product Code. 15868267 Shop All DuPont Products
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15868267
Brand: DuPont™ D15482751
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Tychem™ 6000 FR – Triple Threat Protection in 1 GarmentA single-layer sleeved-apron offering triple-hazard protection from liquid-chemical splash, flash fire, and electric arc.
  • DuPont™ Tychem™ ThermoPro Apron. Sleeved apron available in bright orange. Adjustable FR buckles at back of waist and shoulder. Integral sleeves with elasticated wrists. Sewn with DuPont™ Nomex™ thread
    Tychem™ ThermoPro accessories provide partial body protection and must be used in conjunction with primary flame resistance clothing that is rated for the fire/arc hazard.
    Tychem™ ThermoPro provides triple hazard protection from liquid-chemical splash, flash fire and electric arc in a single layer. They combine the trusted chemical protection of Tychem™ and thermal/arc protection of Nomex™ to help provide industrial workers and HazMat responders with permeation protection against a broad range of toxic industrial chemicals and to provide escape time from flash fires. Ideal for laboratories or for use on top of an existing flame resistant (FR) garments where extra chemical protection is required.
  • Partial body chemical protective clothing, Category III, Type PB [3]
  • Inherently antistatic (EN 1149-5) - on inside
  • EN ISO 11612 (heat and flame), EN ISO 14116 (limited flame spread), IEC 61482-2 (electric arc)
  • Stitched and over-taped seams with barrier tape for protection and strength
Product Title
DuPont™ Tychem™ 6000 FR ThermoPro >