Learn More
Saint Gobain Life Sciences Tygon™ 3350 Sanitary Silicone Tubing
Designed to reduce the risk of particle entrapment and microscopic build-up during fluid transfer. Tygon™ 3350 Sanitary Silicone Tubing features an ultra-smooth inner surface and reduced surface area for improved flow characteristics.
108.00 € - 1095.00 €
Specifications
Product Type | Tubing |
---|---|
Overall Length (English) | 49.2 ft. |
Overall Length (Metric) | 15 m |
Min. Operating Temperature (English) | -112°F |
Min. Operating Temperature (Metric) | -80°C |
Product Code | Brand | Diameter (Metric) Inner | Diameter (Metric) Outer | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inner | Diameter (Metric) Outer | Price | Quantity & Availability | ||||
12356899
|
Saint Gobain Life Sciences
T330D-153 |
3 mm | 6 mm |
212.00 €
Each |
Estimated Shipment: 05-02-2025
Log in to see stock. |
||||
12376899
|
Saint Gobain Life Sciences
T330F-153 |
4 mm | 7 mm |
250.00 €
Each |
Estimated Shipment: 13-02-2025
Log in to see stock. |
||||
12396899
|
Saint Gobain Life Sciences
T330H-153 |
5 mm | 8 mm |
233.00 €
Each |
Estimated Shipment: 09-10-2024
Log in to see stock. |
||||
12306909
|
Saint Gobain Life Sciences
T330I-153 |
6 mm | 9 mm |
273.00 €
Each |
Estimated Shipment: 13-02-2025
Log in to see stock. |
||||
12346909
|
Saint Gobain Life Sciences
T330M-153 |
8 mm | 11 mm |
680.00 €
Each |
Estimated Shipment: 20-12-2024
Log in to see stock. |
||||
11784648
|
Saint Gobain Life Sciences
T330E-203 |
3 mm | 7 mm |
277.00 €
15m |
Estimated Shipment: 13-02-2025
Log in to see stock. |
||||
12386899
|
Saint Gobain Life Sciences
T330G-203 |
4 mm | 8 mm |
309.00 €
Each |
Estimated Shipment: 13-02-2025
Log in to see stock. |
||||
12316909
|
Saint Gobain Life Sciences
T330J-203 |
6 mm | 10 mm |
378.00 €
Each |
Estimated Shipment: 13-02-2025
Log in to see stock. |
||||
12356909
|
Saint Gobain Life Sciences
T330N-203 |
8 mm | 12 mm |
398.00 €
Each |
Estimated Shipment: 18-11-2024
Log in to see stock. |
||||
12376909
|
Saint Gobain Life Sciences
T330P-203 |
10 mm | 14 mm |
486.00 €
Each |
Estimated Shipment: 18-11-2024
Log in to see stock. |
||||
12326909
|
Saint Gobain Life Sciences
T330K-303 |
6 mm | 12 mm |
550.00 €
Each |
Estimated Shipment: 07-11-2024
Log in to see stock. |
||||
12396909
|
Saint Gobain Life Sciences
T330R-303 |
15 mm | 21 mm |
1040.00 €
Each |
Estimated Shipment: 13-02-2025
Log in to see stock. |
||||
12306919
|
Saint Gobain Life Sciences
T330S-303 |
18 mm | 24 mm |
1095.00 €
Each |
Estimated Shipment: 13-02-2025
Log in to see stock. |
||||
12336899
|
Saint Gobain Life Sciences
T330B-103 |
2 mm | 4 mm |
118.00 €
Each |
Estimated Shipment: 26-11-2024
Log in to see stock. |
||||
12346899
|
Saint Gobain Life Sciences
T330C-103 |
3 mm | 5 mm |
159.00 €
Each |
Estimated Shipment: 02-10-2024
Log in to see stock. |
||||
12326899
|
Saint Gobain Life Sciences
T330A-103 |
1 mm | 3 mm |
108.00 €
Each |
In Stock
Log in to see stock. |
||||
12336909
|
Saint Gobain Life Sciences
T330L-153 |
7 mm | 10 mm |
398.00 €
15m |
Estimated Shipment: 02-10-2024
Log in to see stock. |
||||
12316919
|
Saint Gobain Life Sciences
T330T-353 |
20 mm | 27 mm | ||||||
11759128
|
Saint Gobain Life Sciences
T330O-303 |
8 mm | 14 mm |
680.00 €
15m |
Estimated Shipment: 16-10-2024
Log in to see stock. |
||||
Description
- Ultra-smooth inner bore reduces potential for particle entrapment
- Minimal extractables help maintain fluid integrity
- Documented biocompatibility for sensitive applications
- Excellent fluid flow characteristics
- Fully tested to ISO 10993 standards to facilitate validation process
Specifications
Tubing | |
15 m | |
-80°C |
49.2 ft. | |
-112°F |