missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ Tygon™ E-3603 Lab Tubing

Formulated for resistance to flex fatigue and abrasion and is less permeable than rubber tubing. Tygon™ E-3603 Lab Tubing handles virtually all inorganic chemicals found in the lab.

Brand:  Masterflex™ 06407-87

 View more versions of this product

Product Code. 15187124

  • 295.00 € / 15m
Estimated Shipment: 20-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Bio-based formulation delivers excellent performance
  • Lot-to-lot consistency for reproducible results
  • Non-toxic and non-contaminating
  • Smooth, polished inner wall
  • Slips easily over fittings and grips securely for simple lab set-ups
  • Contains no BPA or phthalates
  • Standard sizes available to hold full vacuum at room temperature
Typical Applications
  • General laboratory
  • Food and beverage
  • Biopharmaceutical
  • Analytical instruments
  • Peristaltic and vacuum pumps
  • Ideal for condensers, incubators, desiccators, gas lines and drain line
Specifications

Specifications

FDA Certified
Autoclavable
9.5 mm
0.59 in.
49.2 ft.
0.09 in.
General laboratory, food and beverage, biopharmaceutical, analytical instruments
Transparent
20 psi
74°C
Laboratory Tubing
0.37 in.
Tygon E-3603
15 mm
15 m
2.4 mm
1 Ea.
Flexible
20 psi
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.