missing translation for 'onlineSavingsMsg'
Learn More

Saint Gobain Life Sciences Tygon™ E-3603 Tubing

Tygon™ E-3603 Tubing for laboratory and vacuum applications.

29.85 € - 790.00 €

Specifications

Color Clear
Product Type Tubing
View More Specs

Products 5
Product Code Brand Diameter (Metric) Outer Price Quantity & Availability  
Product Code Brand Diameter (Metric) Outer Price Quantity & Availability  
15108106
View Documents
Saint Gobain Life Sciences
ACF00012
8 mm
106.00 €
15m
Estimated Shipment: 06-05-2025
to see stock.
Add to basket
 
15690211
View Documents
Saint Gobain Life Sciences
ACF00004
4 mm
29.85 €
15m
Estimated Shipment: 24-04-2025
to see stock.
Add to basket
 
10140121
View Documents
Saint Gobain Life Sciences
ACF00030
22.3 mm
790.00 €
15m
Estimated Shipment: 05-03-2025
to see stock.
Add to basket
 
15610221
View Documents
Saint Gobain Life Sciences
ACF00038
11.1 mm
383.00 €
15m
Estimated Shipment: 12-03-2025
to see stock.
Add to basket
 
15517995
View Documents
Saint Gobain Life Sciences
ACF00062
31.8 mm
636.00 €
15m
Estimated Shipment: 17-04-2025
to see stock.
Add to basket
 
Description

Description

Tygon™ E-3603 tubing from Saint-Gobain is crystal clear and flexible, handling virtually all organic chemicals found in the modern laboratory. It is non-oxidizing and non-contaminating, making it an ideal choice for a variety of applications. Tygon™ E-3603 has high tolerances and lot-to-lot consistency, which will yield reproducible results in your laboratory. It is non-toxic and non-contaminating, and less permeable than rubber tubing. The glassy-smooth inner bore helps prevent buildup to facilitate cleaning.
  • Lot-to-lot consistency for reproducible results
  • Non-toxic and non-contaminating
  • Smooth, polished inner wall
  • Slips easily over fittings and grips securely for simple lab set-ups
  • Contains no BPA or phthalates
  • Standard sizes available to hold full vacuum at room temperature
Specifications

Specifications

Clear
Tubing
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.