Learn More
Masterflex™ Tygon™ 2 Stop Hydrocarbon Tubing
Tailored for petro-chemicals and resists swelling and cracking. Tygon™ 2 Stop Hydrocarbon Tubing is also UV and Ozone resistant.
Brand:  Masterflex™ 95606-40
 View more versions of this product
Product Code. 11927875
  • 62.75 € / Pack of 12
Estimated Shipment: 10-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Resists embrittlement
  • Compatible with most hydrocarbons
  • Resistant to swelling
  • Highly flexible, easy to install
  • Specifically developed for fuels and lubricants
  • Ozone and UV light resistant
Specifications
Specifications
Green/Green
Tygon F4040
0.85 mm
1.85 mm
381 mm
12 Pack