missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ismatec™ Tygon MHSL 2001 Standard Tubing
Transparent, plasticizer-free tubing with superior pump life. Ismatec™ Tygon™ MHSL 2001 Standard Tubing is especially designed for MEK and other aggressive solvents.
Brand: Ismatec™ SC0830
This item is not returnable.
View return policy
Description
- Plasticizer and oil-free.
- Smooth inner-bore.
- Low sorption maintains fluid and tube integrity.
- Does not impart anything into the pumping medium.
- No release of hazardous materials when properly incinerated.
- Applications
- Acids: Excellent
- Alkaline solutions: Excellent
- Solvents: Good/Excellent
- Complies with: FDA.
- Sterilization
- Autoclavable with steam, 30 minutes at 1 bar (15 psi) and 121°C.
- Gas sterilization with ethylene oxide.
- Sterilization with radiation up to 2.5 mrad.
Specifications
FDA | |
Autoclavable | |
Tygon MHSL 2001 | |
15 m | |
peristaltic pump | |
-73°C | |
Acids, Alkalies | |
CO2 1140, O2 76, N2 190 | |
57°C |
Standard Tubing | |
1.6 mm | |
4.8 mm | |
1.6 mm | |
MEK and other aggressive solvents. | |
1 Each | |
1.6 mm I.D., 4.8mm O.D. | |
Transparent |