Learn More
Masterflex™ Tygon™ R-3603 Lab Tubing
Formulated for resistance to flex fatigue and abrasion and is less permeable than rubber tubing. Tygon™ R-3603 Lab Tubing handles virtually all inorganic chemicals found in the lab.
Brand:  Masterflex™ ACF00005-C
 View more versions of this product
Product Code. 10160551
  • 66.45 € / 15m
Estimated Shipment: 21-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Bio-based formulation delivers excellent performance
  • Lot-to-lot consistency for reproducible results
  • Non-toxic and non-contaminating
  • Smooth, polished inner wall
  • Slips easily over fittings and grips securely for simple lab set-ups
  • Contains no BPA or phthalates
  • Standard sizes available to hold full vacuum at room temperature
Typical Applications
  • General laboratory
  • Food and beverage
  • Biopharmaceutical
  • Analytical instruments
  • Peristaltic and vacuum pumps
  • Ideal for condensers, incubators, desiccators, gas lines and drain line
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FDA Compliant, USP Class VI, NSF 51
2.4 mm
0.22 in.
49.2 ft.
0.06 in.
425%
74°C
Sterile
56 (Shore A, 15 sec)
20 psi
74°C
0.09 in.
Plastic
5.6 mm
15 m
1.6 mm
General laboratory, food and beverage, biopharmaceutical, analytical instruments
1 Ea.
Transparent
Flexible
760 mm Hg
Tubing