missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Type 90 Safety Storage Cabinet S-PHOENIX Touchless-90 Light Gray
Safe and approved storage of flammable hazardous substances in working areas
Brand: asecos™ 3012000133719
Alert:
For indoor use only
Description
- Robust construction and longevity: triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
- Space saving folding doors: less required space in front of the cabinet while doors are open
- Effortless use: the sensor detects a person within approx. 2 seconds
- User-friendly and safe: after detection, the folding doors open automatically within approx. 5 seconds. The sensor recognises the person while operating and hold the folding doors open
- Always closed: if the person leaves the sensor-controlled detection area, the folding doors close automatically after approx. 30 seconds
- No unauthorised use: doors lockable with profile cylinder (integration in an existing locking system possible)
- Easy alignment: adjusting aids to compensate for uneven floor
- Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system
Specifications
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, CE compliant | |
616 mm | |
524 x 1050 x 1742 mm | |
Folding, Automatic opening | |
1968 mm | |
S90.196.120.FDAO | |
3 | |
Powder-coated plain | |
1196 mm | |
Light Gray | |
Safety storage cabinet |
3 x tray shelf, 1 x perforated insert (365 mm), 1 x bottom collecting sump (sheet steel powder-coated) | |
524 mm | |
616 x 1196 x 1968 mm | |
Safe and approved storage of flammable hazardous substances in working areas | |
1742 mm | |
2 | |
Tray shelf | |
479.50 kg | |
1050 mm | |
Steel |