Learn More
Tyramine, 98+%, Thermo Scientific Chemicals
Reagent for fluorescence enzyme immunoassay
30.55 € - 130.65 €
Chemical Identifiers
CAS | 51-67-2 |
---|---|
Molecular Formula | C8H11NO |
Molecular Weight (g/mol) | 137.182 |
MDL Number | MFCD00008193 |
InChI Key | DZGWFCGJZKJUFP-UHFFFAOYSA-N |
Synonym | tyramine, 4-2-aminoethyl phenol, 4-hydroxyphenethylamine, p-tyramine, 2-4-hydroxyphenyl ethylamine, uteramine, tyramin, tyrosamine, tocosine, 4-hydroxyphenylethylamine |
PubChem CID | 5610 |
ChEBI | CHEBI:15760 |
IUPAC Name | 4-(2-aminoethyl)phenol |
SMILES | C1=CC(=CC=C1CCN)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15403669
|
Thermo Scientific Alfa Aesar
J60990.03 |
1 g |
30.55 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15413669
|
Thermo Scientific Alfa Aesar
J60990.06 |
5 g |
38.59 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15423669
|
Thermo Scientific Alfa Aesar
J60990.14 |
25 g |
130.65 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tyramine acts as a adrenergic and It is primarily used an intermediate in the manufacture of pharmaceutical and agrochemical products. It Is a vaso active aminoacid found in foods and it It precipitates headaches in some sufferers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTyramine acts as a adrenergic and It is primarily used an intermediate in the manufacture of pharmaceutical and agrochemical products. It Is a vaso active aminoacid found in foods and it It precipitates headaches in some sufferers.
Solubility
Soluble in water (10 mg/ml at 15°C), methanol, DMSO, boiling alcohol (100 mg/ml), and ethanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong acids, strong oxidizing agents.
Chemical Identifiers
51-67-2 | |
137.182 | |
DZGWFCGJZKJUFP-UHFFFAOYSA-N | |
5610 | |
4-(2-aminoethyl)phenol |
C8H11NO | |
MFCD00008193 | |
tyramine, 4-2-aminoethyl phenol, 4-hydroxyphenethylamine, p-tyramine, 2-4-hydroxyphenyl ethylamine, uteramine, tyramin, tyrosamine, tocosine, 4-hydroxyphenylethylamine | |
CHEBI:15760 | |
C1=CC(=CC=C1CCN)O |
Specifications
51-67-2 | |
White to Yellow | |
C8H11NO | |
1099914 | |
tyramine, 4-2-aminoethyl phenol, 4-hydroxyphenethylamine, p-tyramine, 2-4-hydroxyphenyl ethylamine, uteramine, tyramin, tyrosamine, tocosine, 4-hydroxyphenylethylamine | |
DZGWFCGJZKJUFP-UHFFFAOYSA-N | |
4-(2-aminoethyl)phenol | |
5610 | |
137.18 | |
Powder |
160°C to 162°C | |
175°C to 181°C (8 mmHg) Hg | |
MFCD00008193 | |
14,9835 | |
Soluble in water (10mg/ml at 15°C),methanol,DMSO,boiling alcohol (100mg/ml),and ethanol. | |
C1=CC(=CC=C1CCN)O | |
137.182 | |
CHEBI:15760 | |
≥98% | |
Tyramine |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P363-P501c
H314-H335
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: III
missing translation for 'einecsNumber' : 200-115-8
missing translation for 'rtecsNumber' : SJ5950000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only