missing translation for 'onlineSavingsMsg'
Learn More

Tyramine hydrochloride, 98%, Thermo Scientific Chemicals

Product Code. 11484893
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Reagent for fluorescence enzyme immunoassay

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-19-5
Molecular Formula C8H12ClNO
Molecular Weight (g/mol) 173.64
MDL Number MFCD00012901
InChI Key RNISDHSYKZAWOK-UHFFFAOYSA-N
Synonym tyramine hydrochloride, 4-2-aminoethyl phenol hydrochloride, 4-hydroxyphenethylamine hydrochloride, tyrosamine hydrochloride, tyramine monochloride, p-tyramine hydrochloride, phenol, 4-2-aminoethyl-, hydrochloride, p-tryamine hydrochloride, tyramine hcl, unii-z5kdh3h147
PubChem CID 66449
IUPAC Name 4-(2-aminoethyl)phenol;hydrochloride
SMILES [H+].[Cl-].NCCC1=CC=C(O)C=C1

Specifications

Melting Point 271°C to 274°C
Quantity 5 g
Beilstein 3627058
Merck Index 14,9835
Formula Weight 173.65
Percent Purity 98%
Chemical Name or Material Tyramine hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'