missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Tyrphostin AG-213, 97%

Brand:  Thermo Scientific™ J67345.#0

 View more versions of this product

Product Code. 15458709

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

5 mg
Ambient temperatures
122520-86-9
220.25g/mol
Solid
Orange
97%
C10H8N2O2S
RG-50864; Tyrphostin 47
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers