missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Tyrphostin AG-213, 97%
Brand: Thermo Scientific™ J67345.MA
This item is not returnable.
View return policy
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
10 mg | |
Ambient temperatures | |
122520-86-9 | |
220.25g/mol | |
Solid |
Orange | |
97% | |
C10H8N2O2S | |
RG-50864; Tyrphostin 47 |