Learn More
DuPont™ Tyvek 500 Laboratory Coats press studs
Offers enhanced protection for body parts that are more exposed to hazardous substances. DuPont™ Tyvek™ 500 Lab Coat with Collar provide an excellent barrier against fine particles and fibres, is ultra-low-linting and antistatically treated.
Brand:  DuPont™ D13396069
Product Code. 12335659
Description
Description
  • Silicon non-added
  • Stitched internal seams
  • 3 pockets and 5 press stud closures
  • Composed of flash spun high density polyethylene
  • Provides an idea balance of protection, durability and comfort
  • Permeable to both air and water vapor, yet repels water-based liquids and aerosols
  • Applications include pharmaceutical handling, chemical protection, lead and asbestos abatement/remediation, general maintenance/operations, spray painting and general clean-up
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Large
Single-use
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
DuPont™ Tyvek 500 Laboratory Coats press studs > Size: Large