Learn More
DuPont™ Tyvek™ 500 Xpert (Eco Pack)
GREENER_CHOICE
Hooded coverall with elasticated waist and ergonomic and protective design.
239.00 € - 923.00 €
Specifications
Gender Unisex
Material Polyethylene
Color White
Ankle Style Elastic
Cuff Style Elastic
View More Specs
Products 12
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
17301609
GREENER_CHOICE
View Documents
DuPont™
D15359234
Small
913.00 €
Pack of 100
Estimated Shipment: 29-04-2025
to see stock.
Add to basket
17513266
GREENER_CHOICE
View Documents
DuPont™
D15359234
Small
239.00 €
Pack of 25
In Stock
to see stock.
Edge
Add to basket
17523266
GREENER_CHOICE
View Documents
DuPont™
D15359243
Medium
239.00 €
Pack of 25
In Stock
to see stock.
Edge
Add to basket
17311609
GREENER_CHOICE
View Documents
DuPont™
D15359243
Medium
913.00 €
Pack of 100
Estimated Shipment: 29-04-2025
to see stock.
Add to basket
17533266
View Documents
DuPont™
D15359254
Large
239.00 €
Pack of 25
In Stock
to see stock.
Add to basket
17321609
GREENER_CHOICE
View Documents
DuPont™
D15359254
Large
913.00 €
Pack of 100
Estimated Shipment: 29-04-2025
to see stock.
Add to basket
17543266
GREENER_CHOICE
View Documents
DuPont™
D15359261
X-Large
239.00 €
Pack of 25
In Stock
to see stock.
Add to basket
17331609
GREENER_CHOICE
View Documents
DuPont™
D15359261
X-Large
913.00 €
Pack of 100
Estimated Shipment: 29-04-2025
to see stock.
Add to basket
17553266
GREENER_CHOICE
View Documents
DuPont™
D15359276
2X-Large
239.00 €
Pack of 25
In Stock
to see stock.
Edge
Add to basket
17341609
GREENER_CHOICE
View Documents
DuPont™
D15359276
2X-Large
913.00 €
Pack of 100
Estimated Shipment: 29-04-2025
to see stock.
Add to basket
17563266
GREENER_CHOICE
View Documents
DuPont™
D15359284
3X-Large
252.00 €
Pack of 25
Estimated Shipment: 29-04-2025
to see stock.
Add to basket
17351609
GREENER_CHOICE
View Documents
DuPont™
D15359284
3X-Large
923.00 €
Pack of 100
Estimated Shipment: 29-04-2025
to see stock.
Add to basket
Description
Description
Hooded coverall is robust yet lightweight at less than 180 grams per garment. Its 3-piece hood provides an optimal fit to the head and face when turning. Tyvek™ garments are composed of flash spun high density polyethylene, providing an ideal balance of protection, durability and comfort. Tyvek™ is permeable to both air and water vapor, yet repels water-based liquids and aerosols. It offers an excellent barrier against fine particles and fibers (down to 1 micron in size), is ultra-low-linting and antistatically treated.
Applications for Tyvek™ 500 Xpert garments include those found in pharmaceutical handling, chemical processing, the oil and gas industry, general maintenance and operations, automatic spray painting and more.
  • No silicone added
  • Elasticated face, wrists and ankles
  • Glued-in waist elastic
  • Ample crotch area for freedom of movement
  • Large, easy-to-grasp zipper pull
  • Tyvek™ auto-lock zipper and zipper flap for increased protection
  • Chemical protective clothing Category III, Type 5-B and 6-B
  • Protection against radioactive contamination
  • Antistatic treatment on both sides
  • Stitched external seams
  • Very low inward leakage due to optimized design
With Eco Pack, there are 25 garments in a value pack together with one IFU, instead of an individual packaging together with one IFU for each garment unit. The product and its performance remain exactly the same but the changes in packaging, reduce the use of paper and plastic and thus help to better protect the planet.
Specifications
Specifications
Unisex
White
Elastic
Stitched
Category III
Regulation (EU) 2016/425, EN 14126, EN 1073-2, EN 1149-5
<6.3 oz.
Polyethylene
Elastic
Hooded
Non-sterile
Chemicals
No
<180 g
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
DuPont™ Tyvek™ 500 Xpert (Eco Pack)