missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tyvek™ 600 Plus with Socks
Composed of flash spun high density polyethylene. Dupont™ Tyvek™ 600 Plus with Socks is permeable to both air and water vapor, yet repels water-based liquids and aerosols and offers an excellent barrier against fine particles and fibers.
Brand: DuPont™ D13495777
688.89 EUR valid until 2024-09-30
Use promo code "21675" to get your promotional price.
This item is not returnable.
View return policy
Description
- Hooded coverall with integrated socks and additional knee-length boot flap.
- Available in white in sizes S to XXXL.
- Robust yet lightweight (<250g per garment).
- Self-adhesive chin flap for tight seal of suit to the mask.
- Elasticated face, wrists and ankles as well as glued-in waist elastic.
- Elasticated thumb loops prevent sleeves from riding up.
Specifications
Hooded Coveralls | |
HDPE | |
Single-use | |
Elastic, Attached Socks, Boot Overflaps | |
Hooded | |
Zipper with Self-adhesive Flap and Chin Flap | |
Category III, Type 4-B, 5-B and 6-B | |
(EU) 2016/425, EN 14126, EN 1149-5 | |
For maintenance and dismantling jobs in the nuclear industry, pharmaceutical manufacturing or in research and biosecurity laboratories, medical applications and when exposed to biological hazards | |
80/Pk. |
Small | |
White | |
Elastic | |
Thumb Loop | |
Elastic | |
Stitched, Taped | |
Self-adhesive chin flap | |
84 to 92 cm | |
<250 g |
The user should make the risk assessment to determine the protection required.