missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tyvek™ 600 Plus with Socks

Composed of flash spun high density polyethylene. Dupont™ Tyvek™ 600 Plus with Socks is permeable to both air and water vapor, yet repels water-based liquids and aerosols and offers an excellent barrier against fine particles and fibers.

Brand:  DuPont™ D13495744

201.07 EUR valid until 2025-12-31
Use promo code "24380" to get your promotional price.


 View more versions of this product

Product Code. 12342259

  • 254.00 € / Pack of 20
Estimated Shipment: 28-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Hooded coverall with integrated socks and additional knee-length boot flap.
  • Available in white in sizes S to XXXL.
  • Robust yet lightweight (<250g per garment).
  • Self-adhesive chin flap for tight seal of suit to the mask.
  • Elasticated face, wrists and ankles as well as glued-in waist elastic.
  • Elasticated thumb loops prevent sleeves from riding up.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Hooded Coveralls
HDPE
Single-use
Elastic, Attached Socks, Boot Overflaps
Hooded
Zipper with Self-adhesive Flap and Chin Flap
Non-sterile
Self-adhesive chin flap
124 to 132 cm
For maintenance and dismantling jobs in the nuclear industry, pharmaceutical manufacturing or in research and biosecurity laboratories, medical applications and when exposed to biological hazards
3X-Large
White, Blue
Elastic
Thumb Loop
Elastic
Stitched, Taped
Category III, Type 4-B, 5-B and 6-B
(EU) 2016/425, EN 14126, EN 1149-5
Elasticated thumb loops
80/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

The user should make the risk assessment to determine the protection required.