Learn More
DuPont™ Tyvek™ IsoClean™ Cleanroom Hood
Shop All DuPont Products
Click to view available options
Product Type: Hood
Description
- Bound hood opening for low particle shedding
- Full face opening
- Ties with loops for adjustable fit
- Accessory
Specifications
Specifications
Product Type | Hood |
Size | One Size |
Material | Polyethylene |
Closure Type | Ties with Loops |
Model | IC668B00 |
Seam Type | Bound |
Sterility | Non-sterile |
Certifications/Compliance (Europe) | (EU) 2016/425, EN 14126, ISO Class 6-9 |
For Use With (Application) | Cleanrooms Within The Biotech, Pharmaceutical, Medical Device Manufacturing, Food Processing, Cosmetics Industry, Electronics Industries as Well as in Other Critical or Controlled Environments. |