Learn More
DuPont™ Tyvek™ IsoClean™ Hood with Attached Mask
Click to view available options
Product Type: Hood
Description
- Bound face opening for low particle shedding
- Aseptically folded
- Breathable and low linting
- The accessories are individually double packed
- The attached mask is a blue sterile cleanroom mask made out of pleated polyethylene outer layer having a width of 17.5 cm. Please note that it is not a PPE (e.g. not an FFP1, 2 or 3) mask.
Specifications
Specifications
Product Type | Hood |
Size | One Size |
Closure Type | Ties with Loops |
Model | IC 689 |
Seam Type | Bound |
Sterility | Sterile |
Certifications/Compliance (Europe) | ISO 11137-1, ISO Class 5 |
For Use With (Application) | Cleanrooms Within The Biotech, Pharmaceutical, Medical Device Manufacturing, Food Processing, Cosmetics Industry as Well as in Other Critical or Controlled Environments. |