missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tyvek™ Isoclean™ Hooded Coverall IC 193 B WH DS

Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Description
- Clean-processed and gamma-sterilized
- The hood fits a medical mask and has attached ties
- Provides an inherent barrier to particles, microorganisms and non-hazardous water-based light liquid splash
- Breathable and low linting
- The packaging system serves as a key element for contamination risk reduction when transferring apparel into clean areas
- Typically used in cleanrooms within the biotech, pharmaceutical, medical device manufacturing, food processing, cosmetics industry, as well as in other critical or controlled environments

Specifications
Specifications
Product Type | Cleanroom Coveralls |
Size | Large |
Gender | Unisex |
Material | HDPE |
Color | White |
Disposable | Single-use |
Waist Style | Elastic at Back |
Ankle Style | Attached Boots |
Cuff Style | Elastic |
Closure Type | Zipper with Storm Flap |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
AHS2447145 | Certificate of Sterility | 17/12/2024 | D15504991 |
AHS2445160 | Certificate of Sterility | 11/12/2024 | D15504991 |
AHS2440129 | Certificate of Sterility | 11/12/2024 | D15504991 |
AHS2442140 | Certificate of Sterility | 11/12/2024 | D15504991 |
AHS2442130 | Certificate of Sterility | 11/12/2024 | D15504991 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DuPont™ Tyvek™ Isoclean™ Hooded Coverall IC 193 B WH DS >
Spot an opportunity for improvement?Share a Content Correction