Learn More
DuPont™ Tyvek™ IsoClean™ IC 270B WS Lab Coats, Sterile, Double Bagged
BIOPHARMA
Gamma-sterilized lab coats with bound neck are asepcially folded and feature bound internal seams, tunnelled at wrists, and a front snap closure.
365.00 € - 376.00 €
Specifications
Product Type Controlled Environments Lab Coat
Gender Unisex
Material Tyvek
Color White
Closure Type Snaps
View More Specs
Products 6
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
17899313
BIOPHARMA
View Documents
DuPont™
D15560759
Small
365.00 €
Pack of 30
Estimated Shipment: 20-01-2025
to see stock.
Add to basket
17819323
BIOPHARMA
View Documents
DuPont™
D15560760
Medium
365.00 €
Pack of 30
Estimated Shipment: 20-01-2025
to see stock.
Add to basket
17879323
BIOPHARMA
View Documents
DuPont™
D15560761
Large
365.00 €
Pack of 30
Estimated Shipment: 20-01-2025
to see stock.
Add to basket
17859323
BIOPHARMA
View Documents
DuPont™
D15560762
X-Large
365.00 €
Pack of 30
Estimated Shipment: 20-01-2025
to see stock.
Add to basket
17879313
BIOPHARMA
View Documents
DuPont™
D15560763
365.00 €
Pack of 30
Estimated Shipment: 20-01-2025
to see stock.
Add to basket
17869313
BIOPHARMA
View Documents
DuPont™
D15560764
3X-Large
376.00 €
Pack of 30
Estimated Shipment: 20-01-2025
to see stock.
Add to basket
Description
Description
  • Clean-processed and gamma-sterilized
  • Bound internal seams
  • Tunnelled elastication at wrists
  • Front snap closure for easy donning and doffing
Tyvek™ IsoClean™ delivers an ideal balance of protection, durability and comfort. Made of high density polyethylene using a patented flash spinning process Tyvek™ IsoClean™ provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash. Tyvek™ IsoClean™ is also breathable and low linting.
Tyvek™ IsoClean™ accessories (option code WS) have been sterilized by gamma- irradiation. They are folded to aid aseptic donning and packaged in an ISO class 5 cleanroom. All DuPont™ Tyvek™ IsoClean™ sterile accessories (option code WS) are packed in a dual barrier packaging system, consisting of an inner and outer easy tear cleanroom bag. The packaging system serves as a key element for contamination risk reduction when transferring apparel into clean areas. The accessories are individually packed and grouped together in an outer bag.
Garments and accessories made of sterile Tyvek™ IsoClean™ are typically used in cleanrooms within the hospital pharmacies, biotech, pharmaceutical, medical device manufacturing, food processing, cosmetics industry as well as in other critical or controlled environments.
Specifications
Specifications
Controlled Environments Lab Coat
Tyvek
Snaps
Single-use
Particulates, Light Liquid Splash
Yes
Unisex
White
Elastic
Sterile
None