missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tyvek™ IsoClean™ IC183B TS Coverall, Sterile, Double Bagged
Unhooded gamma-sterilized coveralls are aseptically folded and feature bound internal seams, tunneled elastication at wrists and ankles, elasticated waist at back, Tyvek™ covered elasticated thumb loops, and a storm flap.
363.00 € - 372.00 €
Specifications
Product Type | Cleanroom Coveralls |
---|---|
Gender | Unisex |
Material | Tyvek™ |
Color | White |
Disposable | Single-use |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
17889413
|
DuPont™
D15560732 |
Small |
363.00 €
Pack of 25 |
Estimated Shipment: 05-12-2024
Log in to see stock. |
|||||
17869403
|
DuPont™
D15560733 |
Medium |
363.00 €
Pack of 25 |
Estimated Shipment: 05-12-2024
Log in to see stock. |
|||||
17819413
|
DuPont™
D15560734 |
Large |
363.00 €
Pack of 25 |
Estimated Shipment: 05-12-2024
Log in to see stock. |
|||||
17829413
|
DuPont™
D15560735 |
X-Large |
363.00 €
Pack of 25 |
Estimated Shipment: 05-12-2024
Log in to see stock. |
|||||
17859413
|
DuPont™
D15560736 |
2X-Large |
363.00 €
Pack of 25 |
Estimated Shipment: 05-12-2024
Log in to see stock. |
|||||
17849413
|
DuPont™
D15560737 |
3X-Large |
372.00 €
Pack of 25 |
Estimated Shipment: 05-12-2024
Log in to see stock. |
|||||
Descrizione
- Gamma-sterilized
- Bound internal seams
- Elasticated waist at back for good fit
- Tyvek™ covered elasticated thumb loops
- Tunneled elastication at wrists and ankles
- Front zipper closure
- Storm flap
- Delivers an ideal balance of protection, durability and comfort
- Made of high density polyethylene using a patented flash spinning process
- Provides an inherent barrier to particles, microorganisms, and non-hazardous water-based light liquid splash
- Breathable and low linting
- Sterilized by gamma-irradiation
- Folded to aid aseptic donning and packaged in an ISO class 5 cleanroom
- Packed in a dual barrier packaging system, consisting of an inner and outer easy tear cleanroom bag
- Packaging system serves as a key element for contamination risk reduction when transferring apparel into clean areas
- Typically used in cleanrooms within the hospital pharmacies, biotech, pharmaceutical, medical device manufacturing, food processing, cosmetics industry as well as in other critical or controlled environments
Specifica
Cleanroom Coveralls | |
Tyvek™ | |
Single-use | |
Elastic | |
Collared | |
Zipper | |
Sterile | |
Yes |
Unisex | |
White | |
Elastic | |
Elastic | |
Unhooded | |
Bound | |
Particle | |
D15560 |