Learn More
DuPont™ Tyvek™ IsoClean™ Slip-retardant Shoe Cover
Click to view available options
Size:
Medium
Large
Description
- Gripper™ soles provide enhanced slip-retardance and robustness
- Delivers an ideal balance of protection, durability and comfort li>Suitable for use in GMP class C/D (ISO Class 6-9) clean rooms
Specifications
Specifications
Size | Medium |
Color | White |
Sterility | Non-sterile |
Model | IC451S00 |
Seam Type | Serged |
Certifications/Compliance (Europe) | (EU) 2016/425, EN 14126 |