Learn More
DuPont™ Tyvek 500 Lab Coat with press studs and pockets
Offers enhanced protection for body parts that are more exposed to hazardous substances. DuPont™ Tyvek™ 500 Lab Coat with Collar provide an excellent barrier against fine particles and fibres, is ultra-low-linting and antistatically treated.
Brand: DuPont™ D13396050
293.06 EUR valid until 2025-06-08
Use promo code "24131" to get your promotional price.
Description
- Silicon non-added
- Stitched internal seams
- 3 pockets and 5 press stud closures
- Composed of flash spun high density polyethylene
- Provides an idea balance of protection, durability and comfort
- Permeable to both air and water vapor, yet repels water-based liquids and aerosols
- Applications include pharmaceutical handling, chemical protection, lead and asbestos abatement/remediation, general maintenance/operations, spray painting and general clean-up

Specifications
X-Large | |
White | |
Single-use | |
3 | |
Straight cut | |
Pharmaceutical handling, chemical protection, lead and asbestos abatement/remediation, general maintenance/operations, spray painting and general clean-up. |
HDPE | |
Press Studs | |
Chemicals | |
Stitched (internal) | |
PPE Category, Antistatic treatment (EN 1149-5) - on both sides |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
The user should make the risk assessment to determine the protection required.