missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tyvek™ Lab Coats, Zipper

Product Code. 11925864 Shop All DuPont Products
Large
Click to view available options
Size:
Small
Large
X-Large
2X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Offers enhanced protection for body parts that are more exposed to hazardous substances. Lab coats are composed of flash spun high density polyethylene, providing an idea balance of protection, durability and comfort.

  • Silicon non-added
  • With collar, zipper closure and 2 pockets
  • Permeable to both air and water vapour, yet repels water-based liquids and aerosols
  • Excellent barrier against fine particles and fibres (down to 1 micron in size), is ultra-low-linting and antistatically treated
  • Applications include pharmaceutical handling, chemical protection, lead and asbestos abatement/remediation, general maintenance/operations, spray painting and general clean-up
TRUSTED_SUSTAINABILITY

Specifications

Product Type Chemical Resistant Lab Coat
Size Large
Gender Unisex
Material HDPE
Color White
Closure Type Zipper
Cuff Style Elastic
Disposable Limited-use
Sterility Non-sterile
Resistance Chemicals
No. of Pockets 2
Seam Type Stitched, Internal
Certifications/Compliance Antistatic treatment (EN 1149-5)
For Use With (Application) Pharmaceutical, Lead and Asbestos Abatement/Remediation, General Maintenancce/Operations, Spray Painting, Clean-up
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DuPont™ Tyvek™ Lab Coats, Zipper >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.