Learn More
DuPont™ Tyvek™ Lab Coats, Zipper Product Code.: 12332249
Add to basket
DuPont™ Tyvek™ Lab Coats, Zipper
Product Code. 12332249 Shop All DuPont Products
Click to view available options
Size:
Small
Large
X-Large
2X-Large
Product Code. 12332249
Brand: DuPont™ D13495983
Edge
Add to basket
Add to basket
Offers enhanced protection for body parts that are more exposed to hazardous substances. Lab coats are composed of flash spun high density polyethylene, providing an idea balance of protection, durability and comfort.
  • Silicon non-added
  • With collar, zipper closure and 2 pockets
  • Permeable to both air and water vapour, yet repels water-based liquids and aerosols
  • Excellent barrier against fine particles and fibres (down to 1 micron in size), is ultra-low-linting and antistatically treated
  • Applications include pharmaceutical handling, chemical protection, lead and asbestos abatement/remediation, general maintenance/operations, spray painting and general clean-up
TRUSTED_SUSTAINABILITY
Specifications
Product Type Chemical Resistant Lab Coat
Gender Unisex
Disposable Limited-use
Sterility Non-sterile
Resistance Chemicals
For Use With (Application) Pharmaceutical, Lead and Asbestos Abatement/Remediation, General Maintenancce/Operations, Spray Painting, Clean-up
Product Content Correction
Product Title
DuPont™ Tyvek™ Lab Coats, Zipper >