Learn More
DuPont™ Tyvek™ PL30NP Collared Laboratory Coat
Offers enhanced protection for body parts that are more exposed to hazardous substances. DuPont™ Tyvek™ PL30NP Collared Laboratory Coats are composed of flash spun high density polyethylene, providing an ideal balance of protection, durability and comfort.
Brand: DuPont™ D13495990
55.56 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Silicon non-added
- With 5 press stud closures
- Elasticated cuffs (not tunneled)
- Stitched internal seams
- Permeable to both air and water vapour, yet repels water-based liquids and aerosols
- Excellent barrier against fine particles and fibres (down to 1 micron in size), is ultra-low-linting and antistatically treated
- Applications include pharmaceutical handling, chemical protection, lead and asbestos abatement/remediation, general maintenance/operations, spray painting and general clean-up
Specifications
Medium | |
White | |
Certified according to Regulation (EU) 2016/425, partial body chemical protective clothing, Category III, Type PB [6-B], EN 14126 (barrier to infective agents), anti-static treatment (EN 1149-1) - on both sides |
Flashspun HDPE | |
With collar and press stud closures, without pockets, with elasticated cuffs and stitched internal seams | |
Pharmaceutical handling, chemical protection, lead and asbestos abatement/remediation, general maintenance/operations, spray painting and general clean-up |