missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tychem 2000 C Gown
Click to view available options
Size:
Large/X-Large
Small/Medium
Description
- Shin length
- Front protection with wrap-over rear closure
- Hook and loop neck closure and waist ties
- Stitched and over-taped seams
- Yellow
- Polymeric barrier coating provides permeation barrier protection against a wide range of inorganic chemicals and biological hazards
Specifications
Specifications
Size | Small/Medium |
Gender | Unisex |
No. of Pockets | None |
Style | Wrap-around |
Product Type | General Purpose Gown |
Material | Tychem |
Color | Yellow |
Closure Type | Wrap-around Waist Ties |
Disposable | Single-use |
Sterility | Non-sterile |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction